Pulloannostelija, Dispensette® S
Toimittaja: Brand
4600100 4600101 4600120 4600121 4600130 4600131 4600140 4600141 4600150 4600151 4600160 4600161 4600170 4600171 4600210 4600211 4600220 4600221 4600230 4600231 4600240 4600241 4600290 4600291 4600310 4600311 4600320 4600321 4600330 4600331 4600340 4600341 4600350 4600351 4600360 4600361
613-5225EA 280 EUR
613-5225 613-5226 613-5227 613-5228 613-5229 613-5230 613-5231 613-5233 613-5234 613-5235 613-5236 613-5237 613-5238 613-5239 613-5240 613-5241 613-5242 613-5243 613-5244 613-5245 613-5246 613-5247 613-5249 613-5288 613-5289 613-5290 613-5291 613-5292 613-5293 613-5294 613-5295 613-5296 613-5297 613-5298 613-5299 613-5329
Pulloannostelija, Dispensette® S
Pulloannostelijat
Fixed volume or variable volume, analogue-adjustable or digital easy calibration.
  • With or without recirculation valve
  • Autoclavable at 121 °C
  • Three different models of volume adjustment available: Fixed (fixed volume for standard applications; simple calibration adjustment with supplied tool) and Analogue (analogue volume display, fast volume adjustment; simple calibration adjustment with supplied tool)
  • Digital EASY CALIBRATION: Easy to read digital display; dispensing volume can be set accurately and reproducibly. Easy calibration technique for adjustment