Repetitive pipette, Multipette® E3 / E3x
Toimittaja: EPPENDORF
4987000.010 4987000.029 4987000.371 4987000.380
613-3892EA 815 EUR
613-3892 613-3893 613-5189 613-5190
Repetitive pipette, Multipette® E3 / E3x
Pipetit ja pipetintäyttäjät Toistopipetit
This pipette enables precise pipetting from 1 μl to 50 ml and makes long pipetting series faster and easier. The aspirated liquid can be dispensed up to 100 times without a refill making the Multipette® the ideal tool for filling plates or large series of tubes (e.g., nucleic acid purification kits).
  • To be used with Eppendorf Combitips advanced®: Automatic tip recognition for ease of use
  • Defined dispensing speed increases the reproducibility of results; eight different speed levels allow the optimum speed for each application
  • One button tip ejection