Pipette tips, ART®, SoftFit L™
Toimittaja: Thermo Fisher Scientific
2770 2749 2769 2749-05 2769-05 2779-05 3751-05 3752-05 2080 3742-05-RI 3721-RI 2739-HR 2749-HR 2769-HR 3722-05-HR 3722-05-RI 3752-05-HR 3752-05-RI 3751-RI 2739-05-HR 2739-05-RI 2739-RI 2749-05-HR 2749-05-RI 2749-RI 2769-05-HR 2769-05-RI 2769-RI 2779-05-HR 2779-05-RI 2779-HR 2779-RI 2789 2789-05-HR 2789-05-RI 2789-HR 2789-RI 3721-05-HR 3721-05-RI 3721-HR 3722-HR 3722-RI 3731-05-HR 3731-05-RI 3731-HR 3731-RI 3732-05-HR 3732-05-RI 3732-HR 3732-RI 3741-HR 3741-RI 3742-HR 3742-RI 3751-05-HR 3751-05-RI 3751-HR 3752-HR 3752-RI 3781-05-HR 3781-05-RI 3781-HR 3781-RI 3782-05-HR 3782-05-RI 3782-HR 3782-RI 2779
732-2237EA 94.1 EUR
732-2237 732-2313 732-2315 732-2314 732-2316 732-2318 732-0329 732-0330 732-4431 732-1566 613-1979 732-1195 732-1191 732-1187 613-1866 732-1561 613-1867 732-1568 613-1985 732-1197 732-1196 732-1194 732-1193 732-1192 732-1190 732-1189 732-1188 732-1186 732-1185 732-1184 732-1183 732-1182 732-4433 732-1198 732-1199 732-1211 732-1212 613-1868 732-1560 613-1869 613-1870 613-1980 613-1871 732-1562 613-1872 613-1981 613-1873 732-1563 613-1874 613-1982 613-1875 613-1983 613-1877 613-1984 613-1878 732-1567 613-1879 613-1880 613-1986 613-1881 732-1569 613-1882 613-1987 613-1883 732-1570 613-1884 613-1988 732-4432
Pipette tips, ART®, SoftFit L™
Pipetinkärjet
ART® SoftFit-L™ pipette tips seal when exposed to potential contaminants and are designed for use with the Rainin® LTS LiteTouch pipettes.
  • Transparent PP
  • Certified free of RNase, DNase, DNA and pyrogens
  • Sterile tips: Sterilised by gamma radiation
  • 'Low Retention' tips: Extremely hydrophobic surface for lower retention, superior precision and measurement reproducibility
Ideal for the pipetting of radioactive samples, nucleic amplification procedures, or any applications where critical sample handling is required.