Super elastic plunger microlitre syringes
Toimittaja: Hospidex
EX-MSE05 EX-MSE10 EX-MSNE05 EX-MSNE10 EX-MSE05R EX-MSNE05R EX-MSNE10R EX-MSE05WTS EX-MSE10WTS EX-MSNE05WTS EX-MSNE10WTS
613-1700EA 64.7 EUR
613-1700 613-1701 613-1702 613-1703 613-1704 613-1706 613-1707 613-1708 613-1709 613-1710 613-1711
Super elastic plunger microlitre syringes
Ruiskut Ruiskut kromatografiaan
Super elastic plunger is made of special Ti-Ni alloy which has superior flexibility and it does not have any problem with friction damage caused by plunger bending. It simply springs back, if the plunger is bent.
  • Superior chemical resistance
  • Plunger can be wound into a coil of 16 mm Ø
  • Accuracy of injection is about ±1%
  • Provided with 3-coloured index labels