VWR® Sealpette™ Pro Single Channel Pipettes, Mechanical, Variable Volume
Toimittaja: JENCONS SCIENTIFIC
480-044 480-039 480-097 480-098
613-1244EA 0 EUR
613-1244 613-0919 613-0922 613-0923
VWR® Sealpette™ Pro Single Channel Pipettes, Mechanical, Variable Volume
Pipetit ja pipetintäyttäjät Pipetit
Robust, lightweight pipettes for both left- and right-handed users, featuring very low tip ejection and pipetting forces.
  • Thermal insulation of internal components
  • Fast and light volume adjustment with small increments and volume locking button
  • Clear and easy to read display
  • Personal identification with an ID tag
  • Easy maintenance with one multi-purpose tool (calibration, opening tool for ID window or tubes)
  • Fully autoclavable
The Sealpette™ tip cone is supplied with a Standard filter for protection against pipette contamination by aerosol or over-More Product Information