96-Well format pipetting device, Vaccu-Pette SP Bel-Art
Toimittaja: Bel-Art Products, a Part of SP
F378760000 F378760001
612-6020EA 153 EUR
612-6020 612-6021
96-Well format pipetting device, Vaccu-Pette SP Bel-Art
Kuoppalevyannostelijat
Vaccu-Pette 96-well pipetting device can be used to aspirate or dispense 96 wells in less than 10 seconds, speeding up repetitive washing, dispensing or aspirating of wells by working simultaneously.
  • Transparent plastic construction
  • Available in sterile/disposable or autoclavable/reusable models
  • Ideal for efficient washing, dispensing, or aspirating of wells
The device speeds up repetitive washing, dispensing or aspirating of wells by working simultaneously. It has a vacuum outlet at one end, and a short length of 3,2 mm internal diameter tubing isMore Product Information