VWR®, Mixing Measuring Cylinders, Tall Form, Class A
Toimittaja: VWR Collection
612-5771EA 44.4 EUR
612-5771 612-5772 612-5773 612-5774 612-5775 612-5776 612-5777
VWR®, Mixing Measuring Cylinders, Tall Form, Class A
Mittalasit Measuring Cylinders
Borosilicate 3.3 glass, class A.
  • Calibrated to contain (TC, In)
  • Hexagonal glass base, NS ground and PE stopper
  • Ring marks at major graduations, marks and inscriptions in blue enamel
Sertifikaatit: DIN EN ISO 4788
Tilaustiedot: Batch certificate included.