VWR®, Spreaders, T-Shaped, Standard Line
Toimittaja: VWR Collection
612-5498EA 182 EUR
612-5498 612-5499
VWR®, Spreaders, T-Shaped, Standard Line
Viljelysauvat mikrobiologiaan Viljelysauva
These PS, sterile, disposable T-shaped spreaders are designed for spreading and dispersing liquids across the surface of agar culture plates. Their design ensures that even pressure is applied across the entire length of the T-bar in contact with the agar surface.
  • Smooth spreading surface
  • Pre-sterilised, eliminating the need for flaming or autoclaving
  • Available individually packed or bulk packed (5 pieces per bag)