Measuring cylinders, tall form, class B, SAN
Toimittaja: VITLAB
65091 65191 64991 65291 64891
612-4311EA 21.6 EUR
612-4311 612-4313 VITL64991 VITL65291 VITL64891
Measuring cylinders, tall form, class B, SAN
Mittalasit Measuring Cylinders
SAN, tall form, class B.
  • Calibrated to contain 'ln'
  • Hexagonal base with bottom studs provides high stability, spout
  • Raised scale and ring marks at the primary scale points
  • Thermal stress up to 60 °C does not cause tolerance limits to be permanently exceeded
Sertifikaatit: DIN 12681 / ISO 6706