Mittapullo, luokka A, with 3 marks, DAkkS-calibrated, BLAUBRAND®
Toimittaja: Brand
38204 38206 38208 38210
612-3656EA 202 EUR
612-3656 612-3657 612-3658 612-3659
Mittapullo, luokka A, with 3 marks, DAkkS-calibrated, BLAUBRAND®
Kolvit ja erlenmeyerit Mittapullot
Borosilicate glass 3.3, with PP stopper.
  • Used to check the functioning of a dispenser
  • Calibrated to contain (TC, In), incl. DAkkS-certificate
  • Marks and inscriptions in high contrast blue enamel
  • DE-M marking
Sertifikaatit: DIN EN ISO 1042