Pipettitelineet, pyörivä
Toimittaja: KARTELL
262 265
612-2814EA 44.1 EUR
612-2814 612-1811
Pipettitelineet, pyörivä
Pipettitelineet
PP, with circular PVC base (both references to be ordered together to have the complete system).
  • For 18 pipettes Ø <10 mm and 10 pipettes Ø <15 mm
  • The base on which the pipettes rest is provided with small drainage holes
  • With fixing ring for height adjustment on shaft of circular base