VWR®, Spreaders, L-shaped, Disposable
Toimittaja: VWR Collection
612-1560EA 162 EUR
612-1560 612-1561
VWR®, Spreaders, L-shaped, Disposable
Viljelysauvat mikrobiologiaan Viljelysauva
PS, sterile. The foot of the spreader has a completely smooth, rounded surface, free of rough edges and imperfections. It enables even spreading of liquid samples across the surface of agar plates without gouging or cutting the medium.
  • Surface treated to enhance adhesion of droplets
  • Non toxic
  • Gamma irradiated
Dimensions: total length 145 mm (±1 mm); spreader length 38 mm (±0,5 mm).