Graduated pipettes, with piston, FORTUNA®
Toimittaja: Poulten & Graf
1.1424702 1.1423204
612-1007EA 101 EUR
612-1007 612-1505
Graduated pipettes, with piston, FORTUNA®
Pipetit ja pipetintäyttäjät Pipetit, asteikolla
Clear glass.
  • Blue or amber graduations
  • Clear calibrated cylinders
  • Tolerance indications
  • Use of pipetting aid not necessary
  • Single pack
Without certificate.