Weighing bottles, PP
Toimittaja: KARTELL
340 342 343 345 346 347 348
611-0103EA 9.8 EUR
611-0103 611-9402 611-0104 611-9403 611-9404 611-9405 611-0105
Weighing bottles, PP
Pullot Punnituslasit
PP with lid.
  • Very good chemical resistance
  • Very good temperature resistance
Cylindrical shape, with knobbed lid.