Moisture Analyser, MB32
Toimittaja: OHAUS
30971922
611-5848EA 2190 EUR
611-5848
Moisture Analyser, MB32
Analysaattorit Moisture Analysers
Perfect for routine moisture testing.
  • An LCD with second line display, navigation knob, and intuitive operation system makes for convenient routine moisture testing
  • The new carbon fiber heating technology and upgraded loadcell shortens analysis time, saves power and keeps results accurate and consistent
  • Robust, slim design with a strong housing ensures MB32's durability even in harsh environments
  • External calibration
The MB32 features a redesigned housing with stronger material, innovative carbon fiber heating technology and an upgraded load cell forMore Product Information