Precision balances, PR series
Toimittaja: OHAUS
30430077 30430071 30492730 30524386 30492736 30430072 30492731 30524387 30430073 30492732 30524608 30430074 30492733 30524609 30430075 30492734 30524610 30430076 30492735 30524611 30524612 30430078 30498176 30524613
611-5009EA 846 EUR
611-5009 611-5022 611-5007 611-4963 611-4967 611-5021 611-4972 611-4962 611-5013 611-4971 611-4961 611-5012 611-4970 611-4960 611-5011 611-4969 611-4959 611-5010 611-4968 611-4958 611-4957 611-5008 611-4966 611-4956
Precision balances, PR series
Vaa'at
The PR series of balances has been designed for routine weighing applications in the workplace.
  • Three essential weighing modes
  • Choice of models with internal or external calibration
  • Balances are easy to operate, with almost no training required
  • Small footprint saves bench space, while providing a large weighing surface
Offering accuracy and repeatability in essential weighing applications in the laboratory, industrial and educational settings, these durable precision balances deliver competitive performance at an economical price. Featuring aMore Product Information