Precision balances, Entris® II Basic Essential series
Toimittaja: Sartorius Balances
BCE1202-1S BCE1202I-1CEU BCE1202I-1S BCE2201-1S BCE2201I-1CEU BCE2201I-1S BCE2202-1S BCE2202I-1CEU BCE2202I-1S BCE223-1S BCE223I-1CEU BCE223I-1S BCE3202-1S BCE3202I-1CEU BCE3202I-1S BCE323-1S BCE323I-1CEU BCE323I-1S BCE4202-1S BCE4202I-1CEU BCE4202I-1S BCE423-1S BCE423I-1CEU BCE423I-1S BCE5201-1S BCE5201I-1CEU BCE5201I-1S BCE6200-1S BCE6200I-1CEU BCE6200I-1S BCE6202-1S BCE6202I-1CEU BCE6202I-1S BCE622-1S BCE622I-1CEU BCE622I-1S BCE623-1S BCE623I-1CEU BCE623I-1S BCE653-1S BCE653I-1CEU BCE653I-1S BCE8200-1S BCE8200I-1CEU BCE8200I-1S BCE8201-1S BCE8201I-1CEU BCE8201I-1S BCE822-1S BCE822I-1CEU BCE822I-1S
611-4560EA 1450 EUR
611-4560 611-4566 611-4568 611-4578 611-4579 611-4586 611-4587 611-4588 611-4590 611-4598 611-4599 611-4605 611-4610 611-4611 611-4613 611-4614 611-4615 611-4617 611-4618 611-4619 611-4621 611-4622 611-4623 611-4625 611-4626 611-4627 611-4629 611-4630 611-4631 611-4633 611-4634 611-4635 611-4637 611-4638 611-4639 611-4641 611-4642 611-4643 611-4645 611-4650 611-4651 611-4653 611-4654 611-4655 611-4657 611-4658 611-4659 611-4661 611-4662 611-4663 611-4665
Precision balances, Entris® II Basic Essential series
Vaa'at
Entris® II Basic Essential precision balances feature isoCAL, LED touch technology screen and 12 built-in applications. Highly accurate results are guaranteed via the monolithic weigh cell technology. High chemical resistance is ensured by using parts made from hard-wearing PBT, stainless steel and glass.
  • Fast stabilisation time thanks to state-of-the-art weighing sensors
  • Best repeatability with rectangular weighing pan
  • Overload protection, rugged design, weighs up to a pre-set amount
  • Guaranteed reliability with the self-test ‘@start’
  • Choice of models with