VWR® TS Professional, Semi-Micro Balances, Touch Screen
Toimittaja: VWR Collection
611-3664EA 2200 EUR
611-3664 611-3666 611-3667 611-3668 611-3669 611-3663 611-3671 611-3672
VWR® TS Professional, Semi-Micro Balances, Touch Screen
Vaa'at
Semi-micro balances that have 5" colour touch screen display with advanced applications and large databases for professional weighing. The user has easy, intuitive access to balance functions, via interactive, icon-driven menus ensuring efficient and productive weighing. Balances can be used by up to 10 different users, each with their own customisable settings and password protection. Ideal for laboratories requiring GLP compliance. All models have a stainless steel top pan.
  • Internal and external calibration (optional factory calibration certificate)