Kannettava vaaka, Scout® SKX -sarja
Toimittaja: OHAUS
30253070 30253071 30253072 30253076 30253073 30253077 30253078 30253074 30253075 30253079 30253080
611-3482EA 600 EUR
611-3482 611-3483 611-3484 611-3488 611-3485 611-3489 611-3490 611-3486 611-3487 611-3491 611-3492
Kannettava vaaka, Scout® SKX -sarja
Vaa'at
The Scout® SKX series has been designed for education applications. The overload protection and large backlit LCD, backed by education software makes the Scout® the ideal balance for the classroom. The integrated transportation lock protects the load cell from damage during stacking storage and transportation. Can be used either in battery-powered mode (4× AA batteries, not included) or via the AC adapter, with power saving auto shut-off function and low battery indicator. The SKX123 has an ingenious draught shield with an easy to remove top cover piece.
  • Overload protection (10