Industrial bench scales, Ranger 4000 series
Toimittaja: OHAUS
30236943 30231099 30231100 30231101 30237016 30237017 30237018 30237019
611-3414EA 773 EUR
611-3414 611-3408 611-3409 611-3410 611-3415 611-3416 611-3417 611-3418
Industrial bench scales, Ranger 4000 series
Vaa'at
Multipurpose, compact scale for producing precise results rapidly. Application modes include weighing, percent weighing, parts counting, accumulation and dynamic weighing and display hold
  • Bright white LED display, suitable for dim light conditions
  • Portable (net weight 5,9 kg) with rugged die-cast metal housing and rechargeable battery
  • Simple keyboard; five key operation and intuitive SmarText™ software
  • Stabilisation within 1 second
  • Checkweighing LEDs and audible indicators
Sertifikaatit: Meets Class III requirements of OIML R76 and EN 45501