Weighing Boats for Micro Balances
Toimittaja: Sartorius Balances
6565-250 6566-50
SARB6565-250EA 119 EUR
SARB6565-250 611-3288
Weighing Boats for Micro Balances
Punnitusastiat Punnitusastiat
Weighing boats made from aluminium. For use with ultra micro and micro balances.