Punnitusalusta, punnituslaiva, PYREX®
Toimittaja: SciLabware
2115/04M 2115/06M 2115/02M
611-0010EA 37.8 EUR
611-0010 611-0011 611-3138
Punnitusalusta, punnituslaiva, PYREX®
Punnitusastiat Punnitusastiat
Borosilicate glass.
  • Ideal for weighing small quantities of powder
  • With hollow stem to facilitate introduction of the powder into narrow neck vessels e.g. volumetric flasks
  • Flat base ensures stability on the balance pan