VWR®, Kannettava vaaka, kannettava välineistö tiheyden mittaamiseen
Toimittaja: VWR Collection
611-3131EA 1350 EUR
611-3131
VWR®, Kannettava vaaka, kannettava välineistö tiheyden mittaamiseen
Vaa'at
A complete portable system for density measurements, comprising of a portable balance plus a density kit for solids and liquids with the required accessories, all presented in a customised carrying case.
  • Precision balance
  • Density kit for solids and liquids
  • Calibration weight class F1 200 g
  • Software installed on the balance for the automatic calibration of solids and liquids
  • Density values are shown directly on the balance display
The LPC-313 portable balance has an optional external battery pack and round draught shield Ø 135 mm to fit the Ø 90&More Product Information