VWR®, Eyepiece camera
Toimittaja: VWR Collection
611-2257EA 184 EUR
611-2257
VWR®, Eyepiece camera
Kamerat
This USB camera fits into the eyepiece tube of biological binocular and monocular microscopes. This enables users to digitise images even when the microscope does not have a trinocular port.
  • For 23 or 30 mm eyepieces
  • Automatic exposure control and white balance
  • Can also be fitted to stereo microscopes
  • Includes software for image analysis and processing
Toimitustiedot: Supplied with USB cable, ring adapters (30 and 30,5 mm), CD ROM with image analysis software and documentation.