Silikageeli, Pore size: 60 Å 35-60 mesh, high purity, Supelco®

Toimittaja: Merck


M-Clarity™ quality level = MQ100

60742-1KG 60742-25KG
60742-1KGEA 148 EUR
60742-1KG 60742-25KG
Silikageeli, Pore size: 60 Å 35-60 mesh, high purity, Supelco®
Silikageeli
Kaava: SiO₂
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011232
CAS-numero: 112926-00-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8ab4cc0b12527585b6641fb374f4805b5c48959&vid=3f8d94354f149f3abd134327fe45819930dc3924&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR