Magnesiumkloridihexahydraatii 99.0-105.0%, lastu FCC, Macron Fine Chemicals™

Toimittaja: Avantor Performance Materials

5954-24EA 636.91 EUR
5954-24 5954-32
Magnesiumkloridihexahydraatii 99.0-105.0%, lastu FCC, Macron Fine Chemicals™
Magnesiumkloridihexahydraatii
Kaava: MgCl₂·6H₂O
Kiehumispiste: 1412 °C (1013 hPa)
Sulamispiste: 117 °C
Tiheys: 1,57 g/cm³ (25 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149781
CAS-numero: 7791-18-6
EINECS: 232-094-6
Merck Index: 13,05684

Order Now

Testitulosten erittely

Food GMP Manufactured Product
Meets F.C.C. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Appearance Passes Test
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Assay (MgCl₂.6H₂O) 99.0 - 105.0 %
FCC - Ammonium (NH₄) ≤0.005 %
FCC - Lead (Pb) ≤4 mg/kg
FCC - Sulfate (SO₄) ≤0.03 %
Solubility Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR