Thread adaptors, TYPE A, b.safe
Toimittaja: Bohlender
M600-22 M600-26 M600-28 M600-29 M600-31 M600-46 M600-49 M600-51 M615-25 M615-27 M615-29 M615-30 M615-32 M615-33 M615-34 M615-35 M615-36 M615-40 M615-41 M615-47 M615-48 M615-50 M615-52 M615-61 M615-63 M615-64 M615-65 M615-75 M630-60 M630-62 M630-66
590-0235EA 93.8 EUR
590-0235 590-0236 590-0237 590-0238 590-0239 590-0240 590-0241 590-0242 590-0248 590-0249 590-0250 590-0251 590-0252 590-0253 590-0254 590-0255 590-0256 590-0257 590-0258 590-0259 590-0260 590-0261 590-0262 590-0263 590-0264 590-0265 590-0266 590-0268 590-0273 590-0274 590-0275
Thread adaptors, TYPE A, b.safe
Kiinnitykset ja liittimet
All adaptors of this type are supplied with a male thread, a female thread and recessed grips.
  • Stay flexible and independent
  • Recessed grips for an easy assembly
Your existing caps and waste caps do not fit with the flasks and canisters which you would like to use or vice versa? Simply use b.safe thread adaptors for a flexible combination of caps and waste caps independently from the thread on the flask and/or container.