Exhaust filters for barrels, b.safe
Toimittaja: Bohlender
M506-21 M506-23 M506-25
590-0179EA 372 EUR
590-0179 590-0180 590-0181
Exhaust filters for barrels, b.safe
Suodattimet Ilmansuodattimet
Filter body made of PE with connection thread for direct installation on barrels. Lid with aeration openings and label field to monitor service life. Filling made of activated carbon for adsorption of solvents.
  • Activation of service life through touch of the button and/or individual marking on the back side
  • Splash-proof ventilation openings
  • Activated carbon in the inside guarantee adsorption of harmful vapours
  • PP for high resistance and fire class rating V-0
  • Plenty of space for fittings and hose connectors
Safe disposal of solvents without leakage ofMore Product Information