Fittings, b.safe
Toimittaja: Bohlender
M501-06 M501-11 M501-12 M501-15 M501-16 M501-17 M501-18
590-0148EA 52.4 EUR
590-0148 590-0149 590-0150 590-0151 590-0152 590-0153 590-0154
Fittings, b.safe
Kiinnitykset ja liittimet
Made of PFA to fix tubing in the capillary connectors (thread UNF ¹/₄") 28G) of b.safe Caps and b.safe Waste Caps. The conical seal cone clamps the tubing and seals the connection safely.
  • Internal cone for comfortable inlet of capillaries
  • Optimized surface design for more grip
  • Centring pin for easy installation
b.safe fittings are available in different colours for all standard HPLC capillaries and their outer diameter. This makes the application clear and simple.