b.safe Waste cap electronic fill-level control
Toimittaja: Bohlender
M202-40 M203-41 M205-40 M205-41 M206-41 M206-42 M206-51 M206-60 M206-61 M208-41 M209-41 M210-41
590-0098EA 642 EUR
590-0098 590-0099 590-0100 590-0101 590-0102 590-0103 590-0104 590-0105 590-0106 590-0107 590-0108 590-0109
b.safe Waste cap electronic fill-level control
Tulpat ja korkit Turvakorkit
To prevent your waste canister from overflowing, connect these b.safe waste caps with electronic fill-level control to the alarm box. When the 'canister full' level is reached, you will receive an optical and acoustic warning. The alarm is triggered before your canister is completely full. This leaves a small safety buffer in the canister.
  • Prevent your waste canisters from overflowing
  • Optical and acoustic warning
  • 2 alarmbox models available
  • Acessories like fittings and hose connectors guarantee you waste lines are connected leak tight
  • Special couplings for