Blotting papers
Toimittaja: Sartorius
FT-2519460570N FT2-521580600G FT2-520580600K FT2-519580600N FT-2519200200N FT2-520200200N FT-3-521-185 FT2520-460570K
588-3851EA 446 EUR
588-3851 515-1632 515-1606 588-3852 SARTFT-2519200200N SARTFT2-520200200N SARTFT-3-521-185 SARTFT2520-460570K
Blotting papers
Blottauspaperit
Their extremely smooth surface and relatively high thickness provide them with superior absorption properties and uniformity across the entire contact area.
  • Grades BF 2 and BF 3 are made of cotton linters with more than 98% alpha-cellulose content
  • Four grades of blotting papers are available
Grades BF 1 and BF 4 are made of refined pulp and cotton linters with more than 95% alpha-cellulose content.