(±)-Taxifolin, hydrate ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®

Toimittaja: Merck Millipore Calbiochem


M-Clarity™ quality level = MQ100

580553-25
580553-25EA 113 EUR
580553-25
(±)-Taxifolin, hydrate ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®
(±)-Taxifolin

An antioxidant flavonoid that scavenges superoxide anion (1.5x105 M-1s-1) and suppresses the cytotoxicity of superoxide ions and hydrogen peroxide in Chinese hamster V79 cells.


Pakkaus: Packaged under inert gas

Kaava: C₁₅H₁₂O₇
Storage Temperature: Jääkaappi
MDL numero: MFCD11044454
CAS-numero: 24198-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR