Supelco® Ascentis® Express AQ-C18 HPLC and UHPLC Columns
Toimittaja: Merck
577329-U 577321-U 565349-U 565351-U 565362-U 565363-U 565364-U 565365-U 577160-U 577320-U 577323-U 577332-U 577335-U 581363-U 582702-U 584572-U 584573-U 584574-U 584575-U 584576-U 584577-U 584578-U 584579-U 584580-U 584581-U 577161-U 577162-U 584582-U 584583-U 584584-U 577338-U 577339-U 577322-U 577324-U 577325-U 577327-U 577328-U 577330-U 577331-U 577333-U 577334-U 577336-U 577337-U 577340-U 577326-U 577341-U 584526-U 577168-U
577329-UEA 835 EUR
577329-U 577321-U 565349-U 565351-U 565362-U 565363-U 565364-U 565365-U 577160-U 577320-U 577323-U 577332-U 577335-U 581363-U 582702-U 584572-U 584573-U 584574-U 584575-U 584576-U 584577-U 584578-U 584579-U 584580-U 584581-U 577161-U 577162-U 584582-U 584583-U 584584-U 577338-U 577339-U 577322-U 577324-U 577325-U 577327-U 577328-U 577330-U 577331-U 577333-U 577334-U 577336-U 577337-U 577340-U 577326-U 577341-U 584526-U 577168-U
Supelco® Ascentis® Express AQ-C18 HPLC and UHPLC Columns
Kromatografiakolonnit
Ascentis® Express AQ-C18 are high-speed, high-performance liquid chromatography columns, based on the Fused-Core® particle design.
The particle provides a thin porous shell of high-purity silica surrounding a solid silica core. This particle design exhibits very high column efficiency due to the shallow diffusion paths in the 0,5 μm thick porous shell and the small overall particle size of 2,7 μm. The AQ-C18 is a C18 bonded phase prepared using a proprietary procedure that incorporates a small amount of polar silane which makes the phase resistant to dewetting. This resistance toMore Product Information