Discovery® HS C18 HPLC Columns
Toimittaja: Merck
568523-U 568500-U 568501-U 568502-U 568503-U 568510-U 568512-U 568513-U 568520-U 568521-U 568522-U 568533-U 568543-U 568572-U 568573-U 568643-U 569250-U 569251-U 569252-U 569253-U 569254-U 569255-U 569274-U 569275-U 569276-U
568523-UEA 921 EUR
568523-U 568500-U 568501-U 568502-U 568503-U 568510-U 568512-U 568513-U 568520-U 568521-U 568522-U 568533-U 568543-U 568572-U 568573-U 568643-U 569250-U 569251-U 569252-U 569253-U 569254-U 569255-U 569274-U 569275-U 569276-U
Discovery® HS C18 HPLC Columns
Kromatografiakolonnit
The Discovery® HS C18 columns provide the best suitable selectivity for many applications.
  • Non-polar, reversed phase column
  • No bleed LC-MS performance
  • Higher hydrophobicity for better resolution of difficult analytes
  • Impurity: <10 ppm metals
  • Surface coverage: 3,2 µmol/m²
Designed to meet the exacting requirements of pharmaceutical analysis and purification. Discovery® columns are also ideal for all application segments requiring reversed phase HPLC.
Tilaustiedot: Column test certificate available inside the box.