Inline PTFE gas filters, Wafergard® MAX
Toimittaja: ENTEGRIS
WGMXMBSS2 WGMXMBRR2 WGMXMBRR4 WGMXMSRR2 WGMXMBSS3 WGMXMBSS4 WGMXMPSS3 WGMXMPRR4 WGMXMPRR2 WGMXMSBW2 WGMXMSBW3 WGMXMSRR4 WGMXMBPRF2
568-0873EA 0 EUR
568-0873 568-0871 568-0872 568-0882 568-0874 568-0875 568-0878 568-0877 568-0876 568-0879 568-0880 568-0883 568-0870
Inline PTFE gas filters, Wafergard® MAX
Suodattimet Kaasusuodattimet
These filters are compatible with high purity gases as well as general nitrogen and CDA applications. Available in Bronze and Silver performance levels, they are specifically designed for high flow and low delta pressure applications and provide ultimate process protection by removing contaminants that could affect process quality.
  • Superior particulate filtration for ultra pure gas system filtration
  • Pre-conditioned for fast start-up (Silver only)
  • Select ultra high purity (Silver) or high purity (Bronze) surface finish
Filter element: Hydrophobic PTFEMore Product Information