J.T.Baker® BAKER ANALYZED®, Buffer Solutions, Colour Coded
Toimittaja: Avantor
5658.0500EA 38.33 EUR
5658.0500 5676.9010 5676.1000 5676.0500 5659.9010 5659.1000 5659.0500 5658.1000 5647.9010 5647.1000 5647.0500 5658.9010 5655-01 5656-01 5657-01
J.T.Baker® BAKER ANALYZED®, Buffer Solutions, Colour Coded
Biokemialliset puskuriliuokset pH-puskuriliuokset
The quality of many BAKER ANALYZED™ reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS).
  • Colour coded
  • Potassium hydrogen phthalate based pH 4 buffer (red)
  • Phosphate based pH 7 buffer (yellow)
  • Borate based pH 10 buffer (blue)
Ready-to-use pH buffers are manufactured in large lots to stringent specifications and utilize quality control procedures to reduce lot-to-lot variability. You can eliminate the time and expense of preparation and standardization of solutions, helping streamline high-volume analytical work in applications such as quality control.
Solutions are available in both clear and colour-coded varieties.
A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Sertifikaatit: Each lot compared to NIST Standard Reference Buffer.