Kleenpak™ Capsules with HDC® II Membrane for Liquid Applications
Toimittaja: Cytiva (Formerly Pall Lab)
KA3J012P6 KA3J045P6 KA3J100P6
561-0002EA 266 EUR
561-0002 561-0003 561-0004
Kleenpak™ Capsules with HDC® II Membrane for Liquid Applications
Suodattimet Membraanisuodatinkapselit
The HDC II filter medium is comprised of a high performance pleated cartridge with a polypropylene filter medium.
  • Clean and robust design
  • Exceptionally simple and reliable operation
  • High security against filter bypass is ensured, even under conditions of pulsed flow
  • High-strength design allows multiple autoclave cycles for extended use and maximum filter economy
  • Polypropylene outer shell is designed to provide long-term assurance of pressure and temperature ratings
  • Internal hold-up volume and dead space is minimized for maximum product recovery
  • A total