Flow controllers
Toimittaja: PARKER HANNIFIN
72-400 72-402 72-428 72-430 72-431
559-0004EA 0 EUR
559-0004 559-0005 559-0006 559-0007 559-0008
Flow controllers
Virtaussäätimet
For regulating and distributing the clean air output from a Parker compressed air dryer, FT-IR purge gas generator or self contained lab gas generator. Manifold and single flow types are available. Manifold types allow clean gas, at the regulated pressure, into the manifold where independently adjustable flow controls may be set to serve three separate instruments. Single flow types include a pressure regulator and a single flow controller. Each flow controller is equipped with a triple scale pressure gauge for 0 - 7 bar (psig, kg/cm²), a pressure regulator and a flow meter mounted on aMore Product Information