Capillary columns, TRACE™ TR-1
Toimittaja: Thermo Fisher Scientific
260A047P 260A395P 260A469P 260A470P 260A130P 260A131P 260A142P 260A143P 260A154P 260A223P 260A241P 260A296P 260A297P 260A308P
555-9352EA 0 EUR
555-9352 555-9265 555-9263 555-9262 555-9350 555-9285 555-9284 555-9283 555-9282 555-9277 555-9273 555-9272 555-9271 555-9269
Capillary columns, TRACE™ TR-1
Kromatografiakolonnit
TRACE™ TR-1 is a general purpose 100% dimethylpolysiloxane non-polar column suitable for a wide range of compounds, including hydrocarbons, esters, pesticides, drugs and amines.
  • Low bleed for increased sensitivity
  • High operating temperature for easy reconditioning
  • Solvent washable