Capillary columns, TraceGOLD™ TG-WaxMS A
Toimittaja: Thermo Fisher Scientific
26087-2860 26087-2970 26087-2980 26087-1420 26087-1300 26087-1430 26087-1440 26087-1540 26087-2230 26087-2240 26087-1310
555-8603EA 0 EUR
555-8603 555-8602 555-8601 555-8923 555-8926 555-8922 555-8921 555-8920 555-8914 555-8913 555-8925
Capillary columns, TraceGOLD™ TG-WaxMS A
Kromatografiakolonnit
Acidic functionality in the polymer structure allows analysis of acidic compounds without derivatisation.
  • Polar phase, acid-deactivated polyethylene glycol
  • Resists oxidative damage and adsorption of acids
  • Excellent peak shapes for high MW acids
  • Equivalent to USP G25, G35 phases