GC injection port septa
Toimittaja: Thermo Fisher Scientific
31303211 31303228 31303230 31303240 313G3211 313G3228 313G3230 313G3240 31303233
555-8241EA 113 EUR
555-8241 555-8243 555-8244 555-8248 555-8193 555-8191 555-8190 555-8188 555-8246
GC injection port septa
Septumi
Quality GC septa for all applications. The low bleed BTO septa are ideal for MS applications and have excellent mechanical properties. The TR-Green septa have a long injection lifetime and low injection port adhesion.
  • Variety of materials and temperature limits for all applications
  • For use in Thermo Scientific™ TRACE™ and FOCUS™ GCs and instruments from other manufacturers
  • Contaminant-free in individual blister packs
Made of low-bleed silicone, these septa have excellent mechanical properties, are ideal for demanding GC-MS applications, and may beMore Product Information