HPLC-kolonni, Hypersil GOLD™ Silica
Toimittaja: Thermo Fisher Scientific
25105-254630 25103-154630 25105-254030 25105-154630 25102-152130 25103-014001 25102-202130 25102-102130 25102-052130
554-4302EA 968 EUR
554-4302 554-4301 555-7590 555-7578 555-7754 554-4475 555-7755 555-7752 554-4300
HPLC-kolonni, Hypersil GOLD™ Silica
Kromatografiakolonnit
This unbonded, highly pure, base-deactivated silica media efficiently separates non polar and moderately polar organic compounds by normal phase chromatography.
  • Outstanding peak shape and sensitivity