HPLC-kolonni, Hypersil GOLD™ HILIC
Toimittaja: Thermo Fisher Scientific
26502-052130 26502-102130 26502-152130 26503-011001 26503-012101 26503-052130 26503-102130 26503-103030 26503-151030 26503-152130 26503-153030 26503-154630 26505-014001 26505-052130 26505-102130 26505-103030 26505-152130 26505-154630 26505-254630
554-4332EA 0 EUR
554-4332 555-6354 555-6356 555-6365 555-6366 555-6375 555-6436 555-6437 555-6440 555-6441 555-6442 554-4333 555-6447 555-6454 555-6459 555-6460 555-6408 555-6411 554-4334
HPLC-kolonni, Hypersil GOLD™ HILIC
Kromatografiakolonnit
Retain highly polar and hydrophilic analytes, which are difficult to analyse with conventional C18 columns.
  • Excellent peak symmetry
  • Narrow peaks for great efficiency
  • Enhanced sensitivity and improved resolution
  • Solutions for high throughput screening