HPLC columns, Hypersil™ ODS C18
Toimittaja: Thermo Fisher Scientific
30105-254630 30105-103030 30105-154030 30105-153030 30105-152130 30105-124630 30105-124030 30105-123030 30105-202130 30105-104030 30105-102130 30105-054630 30105-014001 30105-013001 30105-012101 30103-254630 30105-104630 30105-154630 30110-254630 30103-252130 30105-254030 30105-253030 30105-252130 30105-204630 30105-204030 30103-254030 30103-064630 30103-154630 30103-154030 30103-124630 30103-124030 30103-104630 30103-104030 30103-102130 30103-054630 30103-014001 30103-013001 30103-012101 30103-103030 30105-053030 30105-122130 30105-052130 30103-153030 30103-152130 30103-053030 30105-014006 30105-022106 30105-024006 30110-104630 30110-204630 30110-304030 30110-304630 30105-259070A 30105-259270A
554-5314EA 880 EUR
554-5314 555-6150 555-6217 555-6216 555-6215 555-6213 555-6211 555-6210 555-6220 555-6151 555-6149 555-6146 554-5316 555-6135 555-6134 555-6132 555-6152 555-6218 555-6183 555-6129 555-6226 555-6225 555-6224 555-6223 555-6222 555-6131 555-6170 555-6127 555-6154 554-5313 555-6177 555-6174 555-6173 555-6171 555-6169 554-5315 555-6163 555-6162 555-6172 555-6144 555-6153 555-6143 555-6180 555-6179 555-6167 555-6136 555-6138 555-6140 555-6235 555-6181 555-6186 555-6187 555-0237 555-0238
HPLC columns, Hypersil™ ODS C18
Kromatografiakolonnit
These ODS columns are suitable for non-polar and moderately polar analytes including acids, neutrals and lipophilic compounds. All column phases are endcapped.
  • C18 stationary phase for a broad range of applications