HPLC columns, Hypersil GOLD™ aQ
Toimittaja: Thermo Fisher Scientific
25305-012106 25305-109570A 25305-159070A 25305-159270A 25305-159370A 25305-159570A 25305-259070A 25305-109070A 25305-109270A 25305-109370A 25305-259270A 25305-259370A 25305-259570A
555-7958EA 580 EUR
555-7958 555-0169 555-0170 555-0171 555-0172 555-0173 555-0174 555-0166 555-0167 555-0168 555-0175 555-0176 555-0177
HPLC columns, Hypersil GOLD™ aQ
Kromatografiakolonnit
Hypersil GOLD™ aQ polar endcapped C18 columns provide a controlled interaction mechanism by which polar analytes can be retained and resolved.
  • Controlled secondary interactions ensure optimal peak shape for all analyte types
  • Ideal for the quantitative analysis of trace levels of polar analytes
  • Excellent peak shape
These endcapped, ultrapure, silica-based columns deliver significant reduction in peak tailing using generic gradients with C18 selectivity. With their excellent resolution, efficiency, and sensitivity, Hypersil GOLD columns give youMore Product Information