UV-lamppu ja Katselulaatikko
Toimittaja: CAMAG
022.9055 022.9060 022.9160 040.2000 040.2100
552-0050EA 0 EUR
552-0050 554-6593 554-6592 CAMA040.2000 CAMA040.2100
UV-lamppu ja Katselulaatikko
Valot UV-lamput
These UV lamps with plastic housing are designed for TLC applications.
  • User friendly and versatile
  • Available in two wavelengths: 254/366 nm
  • Automatic switch off timer after 10 minutes reduces the risk of exposure to radiation
  • UV lamp 4: Tilt sensor automatically turns off the lamp in case the lamp is tilted more than 30 degrees
The viewing box is suitable for inspecting thin layer chromatograms and other objects in an undarkened room. The front of the box is closed with a roller shutter which can be pushed open to the left or to the right as requiredMore Product Information