HPLC columns, Cogent C8 300™
Toimittaja: MICROSOLV
40208-05P-2 40208-10P-1 40208-10P-2 40208-15P-1 40208-15P-2 40008-05P 40008-05P-1 40008-05P-2 40008-05P-2-3M 40008-10P 40008-10P-1 40008-10P-2 40008-10P-2-3M 40008-10P-3M 40008-15P 40008-15P-1 40008-15P-2 40008-15P-2-3M 40008-15P-3M 40008-25P 40008-25P-3M 40008-HG5 40008-HG6 40008-P21-100 40008-P21-150 40008-P21-250 40008-P21-50 40008-P30-100 40008-P30-150 40008-P30-250 40008-P30-50 40008-SP100 40008-SP150 40008-SP250 40008-SP250-3M 40008-SP50
554-6437EA 1120 EUR
554-6437 554-6440 554-6441 554-6444 554-6445 554-6102 554-6103 554-6104 554-6105 554-6108 554-6109 554-6110 554-6111 554-6112 554-6115 554-6116 554-6117 554-6118 554-6119 554-6122 554-6126 554-6140 554-6142 554-6149 554-6150 554-6151 554-6152 554-6154 554-6155 554-6156 554-6157 554-6158 554-6159 554-6160 554-6161 554-6162
HPLC columns, Cogent C8 300™
Kromatografiakolonnit
Using proprietary bonding technology, the Cogent C8 300™ has a C8 chain bonded directly to the 5 µm, 300A TYPE-C™ silica surface with two separate points of attachment resulting in silcon-carbon bonds.
  • Extremely long column life
  • Polar and non-polar analytes
  • Increase precision of your results
  • Based on silica hydride
  • Extremely low pH is possible with this column
The bonded does not hydrolyse in acidic conditions since there are no siloxane bonds attaching the ligands. This column is a very stable, efficient and unique HPLC column for macro molecules. AMore Product Information