High pressure tees, crosses and manifold, Upchurch Scientific®
Toimittaja: UPCHURCH SCIENTIFIC
P-170 P-715 P-726 P-727 P-727-01 P-728 P-729 P-730 U-428 U-428-01 U-429-01 U-430 U-431
554-3096EA 309 EUR
554-3096 UPCHP-715 UPCHP-726 554-3170 554-3171 UPCHP-728 554-3172 UPCHP-730 554-3252 554-3253 554-3255 UPCHU-430 554-3256
High pressure tees, crosses and manifold, Upchurch Scientific®
Kiinnitykset ja liittimet
Made of stainless steel or PEEK.
  • Pressure rating up to 20000 psi (1380 bar) for stainless steel tees and crosses
  • Pressure rating up to 3500 psi (241 bar) for PEEK tees and crosses
  • Pressure rating up to 5000 psi (345 bar) for PEEK manifold