Capillary tubing
Toimittaja: Trajan Scientific and Medical
062442 0624431 0624432 0624434 062444 062445 0624450 0624455 062446 0624460 0624461 0624465 0624469 062447 0624470 0624471 0624473 0624474 0624479 062448 062449 062454 062456 062458 062460 0624625 062463 0624655 062466 062469 062470 062472 062473 062475 062476 062478 062479 062481 062482 062492 064034 064050 064052 064054 064061 064062 064102 064108 064114 0624459 064033 0624476 0624463 0624475 0624477 0624478 062457 062461 0624635 0624685 062474 064051 064103 0624491 062713 062712 06244502 06244501 06244503 062710 062714 062711
554-2716EA 53 EUR
554-2716 554-2717 554-2718 554-2719 554-2720 554-2721 554-2722 554-2726 554-2728 554-2729 554-2730 554-2732 554-2733 554-2734 554-2735 554-2736 554-2737 554-2738 554-2742 554-2743 554-2744 554-2746 554-2747 554-2749 554-2763 554-2765 554-2766 554-2768 554-2769 554-2771 554-2772 554-2773 554-2774 554-2776 554-2777 554-2778 554-2779 554-2780 554-2781 554-2782 554-2789 554-2790 554-2792 554-2793 554-2794 554-2795 554-2796 554-2798 554-2799 554-2727 554-2788 554-2740 554-2731 554-2739 554-3491 554-2741 554-2748 554-2764 554-2767 554-2770 554-2775 554-2791 554-2797 554-2745 554-2786 554-2785 554-2724 554-2723 554-2725 SGEA062710 SGEA062714 SGEA062711
Capillary tubing
Letku Chromatography Tubing
Non deactivated fused silica capillary tubing.
Used in a wide range of capillary GC, HPLC and bioanalytical applications
Made from high quality fused silica
Tubing protected with a high temperature polyimide resin
  • Used in a wide range of capillary GC, HPLC and bioanalytical applications
  • Made from high quality fused silica
  • Tubing protected with a high temperature Polyimide resin
Methyl deactivated fused silica capillary tubing.
Chemically inert and thermally stable up to 380 °C
Suitable for organic and aqueous solvents
Quality guaranteed – individuallyMore Product Information