HPLC-kolonni, PRP-3
Toimittaja: HAMILTON BONADUZ
79392 79794 79382 79574 79468 79526 79469 79461 79393 79454 79395 79466 79124 79123 79147 79186 79295 79920
554-0222EA 1110 EUR
554-0222 554-0467 554-0215 554-1316 554-0250 554-0291 554-0251 554-1275 554-1257 554-1269 554-1258 554-0248 554-1224 554-1223 554-2530 554-2532 HAMI79295 HAMI79920
HPLC-kolonni, PRP-3
Kromatografiakolonnit
PRP-3 polymeric reversed phase columns are ideal for gradient separations of peptides and proteins. The columns are packed with a poly(styrene-divinyl)benzene support.
  • Better sample recovery than silica based columns
  • Excellent durability
  • Last longer than silica based columns
  • Crosslinked to prevent shrinkage or swelling
Since the support is entirely polymeric and there are no C18 groups to hydrolyse, column lifetime is longer. Regardless of whether you use ionpair reagents or high percentage aqueous buffers (80 to 100%) for your separation, PRP-3More Product Information